Product Details

SNP ID
rs35094180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40571608 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTACCTATGTCTTTGACAGGACA[A/G]TGATTTCTTGCGGAACACAGTGCAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C15orf57 PubMed Links

Gene Details

Gene
C15orf57
Gene Name
chromosome 15 open reading frame 57
There are no transcripts associated with this gene.

Gene
RPUSD2
Gene Name
RNA pseudouridylate synthase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286407.1 473 Missense Mutation AAT,AGT N143S NP_001273336.1
NM_152260.2 473 Missense Mutation AAT,AGT N204S NP_689473.1

View Full Product Details