Product Details

SNP ID
rs66951403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6727671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAATGTCTTCAACTCTGAATTTAG[G/T]GTAGGAATTGGATGACGTATGGAGC
Phenotype
MIM: 616009
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
COPS7A PubMed Links

Gene Details

Gene
COPS7A
Gene Name
COP9 signalosome subunit 7A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164093.1 Intron NP_001157565.1
NM_001164094.1 Intron NP_001157566.1
NM_001164095.2 Intron NP_001157567.1
NM_016319.3 Intron NP_057403.1
XM_005253694.2 Intron XP_005253751.1

View Full Product Details