Product Details

SNP ID
rs115149182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:21126798 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCGGAATTTCCCTCCGGAAGATGA[G/A]GGCACTGTAGTTATGCGGGTGGACC
Phenotype
MIM: 616160
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
DHRS7B PubMed Links

Gene Details

Gene
DHRS7B
Gene Name
dehydrogenase/reductase 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015510.4 210 Intron NP_056325.2
XM_005256595.4 210 UTR 5 XP_005256652.2
XM_005256596.3 210 Intron XP_005256653.1
XM_011523786.2 210 UTR 5 XP_011522088.1
XM_017024425.1 210 Intron XP_016879914.1
XM_017024426.1 210 UTR 5 XP_016879915.1

View Full Product Details