Product Details

SNP ID
rs148870824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:5825410 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTCATCTAGTGTCCAAGGCCAC[A/G]TGTCCATTTCCTCAGAAGCAGCAGG
Phenotype
MIM: 602462 MIM: 604831
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRMP1 PubMed Links

Gene Details

Gene
CRMP1
Gene Name
collapsin response mediator protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014809.2 Intron NP_001014809.1
NM_001288661.1 Intron NP_001275590.1
NM_001288662.1 Intron NP_001275591.1
NM_001313.4 Intron NP_001304.1
Gene
EVC
Gene Name
EvC ciliary complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306090.1 Intron NP_001293019.1
NM_001306092.1 Intron NP_001293021.1
NM_153717.2 Intron NP_714928.1
XM_006713865.3 Intron XP_006713928.1
XM_006713866.3 Intron XP_006713929.1
XM_011513419.2 Intron XP_011511721.1
XM_017007883.1 Intron XP_016863372.1

View Full Product Details