Product Details
- SNP ID
-
rs73469949
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:89737833 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATCTCCTCCCCACTCTGTGTCCCC[C/G]TCTACCATAGGCCAAAGGGTACCGC
- Phenotype
-
MIM: 608689
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MESP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75510286] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MESP1
- Gene Name
- mesoderm posterior bHLH transcription factor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018670.3 |
|
Intron |
|
|
NP_061140.1 |
- Gene
- WDR93
- Gene Name
- WD repeat domain 93
View Full Product Details