Product Details

SNP ID
rs61732683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:41130994 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCATTGCGCTTCAGAGAAACGCA[C/T]TTCAGCCACATATTGTGGCTAGGAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KBTBD6 PubMed Links

Gene Details

Gene
KBTBD6
Gene Name
kelch repeat and BTB domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152903.4 1807 Silent Mutation AAA,AAG K506K NP_690867.3
Gene
LOC101929140
Gene Name
uncharacterized LOC101929140
There are no transcripts associated with this gene.

View Full Product Details