Product Details

SNP ID
rs17798549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:89329625 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAACAATTCTTAGATTATACCCAA[G/T]GCAGAATGTTTAATGAAGCAGTTGA
Phenotype
MIM: 616175
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
UBE2J1 PubMed Links

Gene Details

Gene
UBE2J1
Gene Name
ubiquitin conjugating enzyme E2 J1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016021.2 1342 UTR 3 NP_057105.2
XM_011535887.1 1342 UTR 3 XP_011534189.1
XM_011535888.2 1342 UTR 3 XP_011534190.1

View Full Product Details