Product Details
- SNP ID
-
rs3803781
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:78993700 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGGCCACAATGATCTGGTCGTCGGT[A/G]TTGGGGATGAACTTGAAGGACGAGA
- Phenotype
-
MIM: 613165
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CANT1
PubMed Links
Gene Details
- Gene
- CANT1
- Gene Name
- calcium activated nucleotidase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001159772.1 |
1474 |
Silent Mutation |
AAC,AAT |
N352N |
NP_001153244.1 |
NM_001159773.1 |
1474 |
Silent Mutation |
AAC,AAT |
N352N |
NP_001153245.1 |
NM_138793.3 |
1474 |
Silent Mutation |
AAC,AAT |
N352N |
NP_620148.1 |
XM_005257021.1 |
1474 |
Silent Mutation |
AAC,AAT |
N352N |
XP_005257078.1 |
XM_005257022.1 |
1474 |
Silent Mutation |
AAC,AAT |
N352N |
XP_005257079.1 |
XM_006721683.1 |
1474 |
Silent Mutation |
AAC,AAT |
N352N |
XP_006721746.1 |
XM_011524291.1 |
1474 |
Silent Mutation |
AAC,AAT |
N352N |
XP_011522593.1 |
XM_011524293.1 |
1474 |
Silent Mutation |
AAC,AAT |
N352N |
XP_011522595.1 |
XM_011524294.1 |
1474 |
Silent Mutation |
AAC,AAT |
N352N |
XP_011522596.1 |
XM_011524295.2 |
1474 |
Silent Mutation |
AAC,AAT |
N352N |
XP_011522597.1 |
View Full Product Details