Product Details
- SNP ID
-
hCV25760374
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:34089652 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGAATATTTGCTGACTGTCTGCTA[C/G]CCTGAATTTCTCCAACACTGAGTTT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DCAF12
PubMed Links
Gene Details
- Gene
- DCAF12
- Gene Name
- DDB1 and CUL4 associated factor 12
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015397.3 |
|
Intron |
|
|
NP_056212.1 |
View Full Product Details