Product Details

SNP ID
hCV25760755
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:6643128 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATAAATGATGTTAAAAAAACTGG[C/G]ATGTGTCTAAACAATAGAGTGCTAT
Phenotype
MIM: 616967
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C17orf100 PubMed Links

Gene Details

Gene
C17orf100
Gene Name
chromosome 17 open reading frame 100
There are no transcripts associated with this gene.

Gene
KIAA0753
Gene Name
KIAA0753
There are no transcripts associated with this gene.

Gene
MED31
Gene Name
mediator complex subunit 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016060.2 662 Intron NP_057144.1
Gene
TXNDC17
Gene Name
thioredoxin domain containing 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032731.3 662 UTR 3 NP_116120.1

View Full Product Details