Product Details

SNP ID
rs113197692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:92593161 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGATTCGGGCGGAGACGAGATT[A/G]GTGATTTGGCGGCTCCGACTGGCGC
Phenotype
MIM: 148760
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF11 PubMed Links

Gene Details

Gene
KIF11
Gene Name
kinesin family member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004523.3 94 UTR 5 NP_004514.2

View Full Product Details