Product Details
- SNP ID
-
hCV25763248
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:70134006 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCCCGAGCTGTTCGAAACTGGTA[A/G]ACAGTTACTGGACGAAGTAGAAGTG
- Phenotype
-
MIM: 300139
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IGBP1
PubMed Links
Gene Details
- Gene
- IGBP1
- Gene Name
- immunoglobulin (CD79A) binding protein 1
There are no transcripts associated with this gene.
View Full Product Details