Product Details
- SNP ID
-
rs6828254
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:109816393 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACGTGTAGTCTGTATGCAGTCTTCA[A/G]TATTTAGCTTATTTGAAGGAGTAGG
- Phenotype
-
MIM: 606468
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GAR1
PubMed Links
Gene Details
- Gene
- GAR1
- Gene Name
- GAR1 ribonucleoprotein
View Full Product Details