Product Details

SNP ID
rs6504233
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:64491665 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGGTTACTACTGGTACTACAACA[G/A]GTACATCAACGTGAAGAAGGGGAGC
Phenotype
MIM: 180630 MIM: 604983
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
DDX5 PubMed Links

Gene Details

Gene
DDX5
Gene Name
DEAD-box helicase 5
There are no transcripts associated with this gene.

Gene
MIR3064
Gene Name
microRNA 3064
There are no transcripts associated with this gene.

Gene
MIR5047
Gene Name
microRNA 5047
There are no transcripts associated with this gene.

Gene
POLG2
Gene Name
DNA polymerase gamma 2, accessory subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007215.3 Intron NP_009146.2
XM_006721651.2 Intron XP_006721714.1
XM_017024072.1 Intron XP_016879561.1

View Full Product Details