Product Details

SNP ID
rs73891757
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:44714584 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACTGCAGTGTTTTCTTCCCTCTG[A/C]CCCCAGCATCCACAACGGGGTGATC
Phenotype
MIM: 609406
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PRR5 PubMed Links

Gene Details

Gene
PRR5
Gene Name
proline rich 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017528.2 Intron NP_001017528.1
NM_001017529.2 Intron NP_001017529.1
NM_001017530.1 Intron NP_001017530.1
NM_001198721.1 Intron NP_001185650.1
NM_015366.3 Intron NP_056181.2
NM_181333.3 Intron NP_851850.1
Gene
PRR5-ARHGAP8
Gene Name
PRR5-ARHGAP8 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181334.5 Intron NP_851851.3

View Full Product Details