Product Details

SNP ID
rs7211875
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:37411381 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAATTAGGGAATGGACCGTTTGGGT[C/T]CCTTTAGCAGTAAGTACAGTGGGAA
Phenotype
MIM: 200350 MIM: 602276
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACACA PubMed Links

Gene Details

Gene
ACACA
Gene Name
acetyl-CoA carboxylase alpha
There are no transcripts associated with this gene.

Gene
TADA2A
Gene Name
transcriptional adaptor 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166105.2 177 Missense Mutation CCC,TCC P6S NP_001159577.2
NM_001291918.1 177 Missense Mutation CCC,TCC P6S NP_001278847.1
NM_001488.4 177 Missense Mutation CCC,TCC P6S NP_001479.4
NM_133439.3 177 Missense Mutation CCC,TCC P6S NP_597683.3
XM_006722044.3 177 UTR 5 XP_006722107.1
XM_011525153.2 177 UTR 5 XP_011523455.1
XM_011525154.2 177 UTR 5 XP_011523456.1
XM_011525155.2 177 UTR 5 XP_011523457.1
XM_017024981.1 177 Missense Mutation CCC,TCC P6S XP_016880470.1
XM_017024982.1 177 Missense Mutation CCC,TCC P6S XP_016880471.1
XM_017024983.1 177 UTR 5 XP_016880472.1
XM_017024984.1 177 UTR 5 XP_016880473.1
XM_017024985.1 177 UTR 5 XP_016880474.1
XM_017024986.1 177 UTR 5 XP_016880475.1

View Full Product Details