Product Details

SNP ID
rs375072478
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:89133238 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGCATTGTGCCTCAAGCACTGC[A/C]AAGCTTCATCCAGACTCACCTTCAT
Phenotype
MIM: 600412 MIM: 612468
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GBP2 PubMed Links

Gene Details

Gene
GBP2
Gene Name
guanylate binding protein 2
There are no transcripts associated with this gene.

Gene
GBP7
Gene Name
guanylate binding protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207398.2 Intron NP_997281.2

View Full Product Details