Product Details

SNP ID
rs79244096
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:100513286 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAATAACTAAAAGAGGGCTTAT[C/T]ATTATTTAAACTTGTGATTCCTTTA
Phenotype
MIM: 614217
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASCC3 PubMed Links

Gene Details

Gene
ASCC3
Gene Name
activating signal cointegrator 1 complex subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284271.1 Intron NP_001271200.1
NM_006828.3 Intron NP_006819.2
NM_022091.4 Intron NP_071374.1
XM_011535394.2 Intron XP_011533696.1
XM_011535395.2 Intron XP_011533697.1
XM_011535396.2 Intron XP_011533698.1
XM_017010205.1 Intron XP_016865694.1
XM_017010206.1 Intron XP_016865695.1

View Full Product Details