Product Details
- SNP ID
-
rs11085804
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:12096253 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGGTTTAAACCTCTATAATTTATT[A/C]TATTAAAGTAACCATTCTATCATAC
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF788
PubMed Links
Gene Details
- Gene
- ZNF788
- Gene Name
- zinc finger family member 788
There are no transcripts associated with this gene.
View Full Product Details