Product Details

SNP ID
rs202059721
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40661875 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTTCTCTTAATCAAATCACATC[C/T]GAGGACTCGATGGGTCGGTTACTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT222 PubMed Links

Gene Details

Gene
KRT222
Gene Name
keratin 222
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152349.2 Intron NP_689562.1
Gene
LOC105371775
Gene Name
uncharacterized LOC105371775
There are no transcripts associated with this gene.

View Full Product Details