Product Details

SNP ID
rs77669071
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:153297851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCATTCAAACCTGAGAAAGGATGG[A/G]CTGGCAGGGGAGGGGGACACAAGGT
Phenotype
MIM: 608197
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PGLYRP3 PubMed Links

Gene Details

Gene
PGLYRP3
Gene Name
peptidoglycan recognition protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052891.2 1613 Intron NP_443123.1
XM_011509118.1 1613 UTR 3 XP_011507420.1
XM_011509119.1 1613 UTR 3 XP_011507421.1
XM_011509120.2 1613 UTR 3 XP_011507422.1

View Full Product Details