Product Details
- SNP ID
-
rs3733103
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:45921103 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AGGGCTTTATTTGAGGACAGAATCA[G/C]CCAGACAAAAGGGCCAGAAAGAGAG
- Phenotype
-
MIM: 607182
MIM: 606568
- Polymorphism
- G/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FYCO1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75928798] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FYCO1
- Gene Name
- FYVE and coiled-coil domain containing 1
- Gene
- LZTFL1
- Gene Name
- leucine zipper transcription factor like 1
There are no transcripts associated with this gene.
View Full Product Details