Product Details

SNP ID
rs3754091
Assay Type
Validated
NCBI dbSNP Submissions
36
Location
Chr.1:224119960 on Build GRCh38
Set Membership
JSNP Validated
Context Sequence [VIC/FAM]
TTTAAGAAACTGTTTTTGAGAACCT[C/A]CAGGGCTGACATCTTTTGGTGGGTG
Phenotype
MIM: 609100
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
FBXO28 PubMed Links

Gene Details

Gene
FBXO28
Gene Name
F-box protein 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136115.2 Intron NP_001129587.1
NM_015176.3 Intron NP_055991.1

View Full Product Details