Product Details

SNP ID
rs3765684
Assay Type
Validated
NCBI dbSNP Submissions
34
Location
Chr.1:101237032 on Build GRCh38
Set Membership
JSNP Validated
Context Sequence [VIC/FAM]
CACTACGCAGTCAGTCGGGGGCAGC[G/A]GCAAGATGCGAAGCGAGCCGTACAG
Phenotype
MIM: 601974
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
LOC101928370 PubMed Links

Gene Details

Gene
LOC101928370
Gene Name
uncharacterized LOC101928370
There are no transcripts associated with this gene.

Gene
S1PR1
Gene Name
sphingosine-1-phosphate receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320730.1 284 Intron NP_001307659.1
NM_001400.4 284 UTR 5 NP_001391.2

View Full Product Details