Product Details

SNP ID
rs3787728
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.21:36071595 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATCCCTATCCTTTTCCCTAAGTCGT[C/T]TGGGACACAGACCCCACCACCCACC
Phenotype
MIM: 114830
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CBR1 PubMed Links

Gene Details

Gene
CBR1
Gene Name
carbonyl reductase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286789.1 Intron NP_001273718.1
NM_001757.3 Intron NP_001748.1
Gene
LOC100133286
Gene Name
uncharacterized LOC100133286
There are no transcripts associated with this gene.

View Full Product Details