Product Details

SNP ID
rs1435166
Assay Type
Validated
NCBI dbSNP Submissions
47
Location
Chr.1:231366564 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
GCATTCCACTGAATTCCTAAGAGTA[T/C]GGACAATATTTCAACTTTGCAAACA
Phenotype
MIM: 606425
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
EGLN1 PubMed Links

Gene Details

Gene
EGLN1
Gene Name
egl-9 family hypoxia inducible factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022051.2 Intron NP_071334.1
XM_005273166.4 Intron XP_005273223.1
XM_005273167.4 Intron XP_005273224.1

View Full Product Details