Product Details

SNP ID
rs8046978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31061588 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGTCCCCCAGCCCCGCCCCTGCT[G/A]CCGGGGCGGCTGAACTCTCACCTGC
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ZNF668 PubMed Links

Gene Details

Gene
ZNF668
Gene Name
zinc finger protein 668
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172668.1 1651 Missense Mutation GCA,GTA A447V NP_001166139.1
NM_001172669.1 1651 Missense Mutation GCA,GTA A470V NP_001166140.1
NM_001172670.1 1651 Missense Mutation GCA,GTA A447V NP_001166141.1
NM_024706.4 1651 Missense Mutation GCA,GTA A447V NP_078982.3

View Full Product Details