Product Details

SNP ID
rs116262037
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:149358247 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTAGGTGGGGTATAGGAGGGCG[A/G]CTGGGTGAGGGAGGGGTCCTCGGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRPEL2 PubMed Links

Gene Details

Gene
GRPEL2
Gene Name
GrpE like 2, mitochondrial
There are no transcripts associated with this gene.

Gene
GRPEL2-AS1
Gene Name
GRPEL2 antisense RNA 1
There are no transcripts associated with this gene.

Gene
PCYOX1L
Gene Name
prenylcysteine oxidase 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301054.1 Intron NP_001287983.1
NM_001301057.1 Intron NP_001287986.1
NM_024028.3 Intron NP_076933.3
XM_011537680.1 Intron XP_011535982.1
XM_011537681.1 Intron XP_011535983.1
XM_011537682.2 Intron XP_011535984.1

View Full Product Details