Product Details

SNP ID
rs201764305
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50246109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGGTCATCTGTGGGGACAGCAGA[A/G]CCCAGCTCCTCATCTACGGACACCT
Phenotype
MIM: 608544 MIM: 602399 MIM: 610053
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HDAC10 PubMed Links

Gene Details

Gene
HDAC10
Gene Name
histone deacetylase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159286.1 Intron NP_001152758.1
NM_032019.5 Intron NP_114408.3
Gene
MAPK12
Gene Name
mitogen-activated protein kinase 12
There are no transcripts associated with this gene.

Gene
TUBGCP6
Gene Name
tubulin gamma complex associated protein 6
There are no transcripts associated with this gene.

View Full Product Details