Product Details

SNP ID
rs61752580
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136739192 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGAGGGTGGCGGCGGCGTGGAGG[A/G]CAGAGGCAAGGCACACGGCGAGGAC
Phenotype
MIM: 612904 MIM: 609379
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LCN10 PubMed Links

Gene Details

Gene
LCN10
Gene Name
lipocalin 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001712.2 992 UTR 3 NP_001001712.2
Gene
LCN6
Gene Name
lipocalin 6
There are no transcripts associated with this gene.

Gene
LOC100128593
Gene Name
uncharacterized LOC100128593
There are no transcripts associated with this gene.

Gene
MIR6722
Gene Name
microRNA 6722
There are no transcripts associated with this gene.

View Full Product Details