Product Details

SNP ID
rs4237768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5986042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGCAGGTGGCATGAGGAGATACC[A/G]TGTGGCCAGAAGAACATGGACATGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR52L1 PubMed Links

Gene Details

Gene
OR52L1
Gene Name
olfactory receptor family 52 subfamily L member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005173.3 944 Missense Mutation CGG,TGG R297W NP_001005173.3

View Full Product Details