Product Details

SNP ID
rs117170603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:100578677 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGACACAGGTGGCCATGGAACT[C/T]GCTGGTAATAGAGGACACATCTCTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GAS2L3 PubMed Links

Gene Details

Gene
GAS2L3
Gene Name
growth arrest specific 2 like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303130.1 Intron NP_001290059.1
NM_001303131.1 Intron NP_001290060.1
NM_174942.2 Intron NP_777602.1
XM_005268804.3 Intron XP_005268861.1
XM_011538219.2 Intron XP_011536521.1
XM_011538220.2 Intron XP_011536522.1
XM_011538221.2 Intron XP_011536523.1
XM_017019206.1 Intron XP_016874695.1
Gene
PIGAP1
Gene Name
phosphatidylinositol glycan anchor biosynthesis class A pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details