Product Details

SNP ID
rs948914646
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:23918755 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGACTTGTCTTTGTTACAAATG[C/T]GTGTGTATATACTGGAAGTACTTGT
Phenotype
MIM: 604496 MIM: 604174
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NKIRAS1 PubMed Links

Gene Details

Gene
NKIRAS1
Gene Name
NFKB inhibitor interacting Ras like 1
There are no transcripts associated with this gene.

Gene
RPL15
Gene Name
ribosomal protein L15
There are no transcripts associated with this gene.

View Full Product Details