Product Details

SNP ID
rs112927852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:87934994 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTCATTATTTTCGCCTTTTCTCT[C/T]CACTCCCTCCTTTCCCGGTTCCTGC
Phenotype
MIM: 603709
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAM22 PubMed Links

Gene Details

Gene
ADAM22
Gene Name
ADAM metallopeptidase domain 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324417.1 225 Intron NP_001311346.1
NM_001324418.1 225 Intron NP_001311347.1
NM_001324419.1 225 Intron NP_001311348.1
NM_001324420.1 225 Intron NP_001311349.1
NM_001324421.1 225 Intron NP_001311350.1
NM_004194.4 225 Intron NP_004185.1
NM_016351.5 225 Intron NP_057435.2
NM_021721.4 225 Intron NP_068367.1
NM_021722.5 225 Intron NP_068368.2
NM_021723.4 225 Intron NP_068369.1
XM_005250445.3 225 Silent Mutation CTC,CTT L70L XP_005250502.1
XM_006716028.3 225 Silent Mutation CTC,CTT L70L XP_006716091.1
XM_006716029.3 225 Silent Mutation CTC,CTT L70L XP_006716092.1
XM_011516318.1 225 Intron XP_011514620.1
XM_011516319.2 225 Intron XP_011514621.1
XM_011516320.2 225 Silent Mutation CTC,CTT L70L XP_011514622.1
XM_011516321.2 225 Silent Mutation CTC,CTT L70L XP_011514623.1
XM_011516322.1 225 Intron XP_011514624.1
XM_011516323.1 225 Intron XP_011514625.1
XM_011516324.1 225 Intron XP_011514626.1
XM_017012329.1 225 Intron XP_016867818.1
XM_017012330.1 225 Intron XP_016867819.1
XM_017012331.1 225 Intron XP_016867820.1
XM_017012332.1 225 Intron XP_016867821.1
XM_017012333.1 225 Intron XP_016867822.1
XM_017012334.1 225 Intron XP_016867823.1
XM_017012335.1 225 Intron XP_016867824.1
XM_017012336.1 225 Intron XP_016867825.1
XM_017012337.1 225 Intron XP_016867826.1
XM_017012338.1 225 Intron XP_016867827.1
XM_017012339.1 225 Intron XP_016867828.1
XM_017012340.1 225 Intron XP_016867829.1
XM_017012341.1 225 Intron XP_016867830.1

View Full Product Details