Product Details

SNP ID
hCV25926420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:112722963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGAGGCCAGCCCGGGTGGGCGG[A/G]AGGGAGCGGGGTCGGGCGTGTTGGG
Phenotype
MIM: 609957
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM19A3 PubMed Links

Gene Details

Gene
FAM19A3
Gene Name
family with sequence similarity 19 member A3, C-C motif chemokine like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004440.1 Intron NP_001004440.1
NM_182759.2 Intron NP_877436.1
Gene
LOC105378910
Gene Name
uncharacterized LOC105378910
There are no transcripts associated with this gene.

Gene
PPM1J
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1J
There are no transcripts associated with this gene.

View Full Product Details