Product Details

SNP ID
rs8005245
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20117883 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGACAGATATGTGGCCATTTGTAA[C/G]CCCCTACACTACATGACCATCATGA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR4K17 PubMed Links

Gene Details

Gene
OR4K17
Gene Name
olfactory receptor family 4 subfamily K member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004715.1 477 Missense Mutation AAC,AAG N159K NP_001004715.1

View Full Product Details