Product Details

SNP ID
rs138913972
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:125707287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTCAGGTTTTTCCAAAACAAATC[A/G]TAAACCCACACCAATTATGTCCAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FLJ37035 PubMed Links

Gene Details

Gene
FLJ37035
Gene Name
uncharacterized LOC399821
There are no transcripts associated with this gene.

Gene
LOC283038
Gene Name
uncharacterized LOC283038
There are no transcripts associated with this gene.

View Full Product Details