Product Details

SNP ID
rs183643597
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:6800727 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGATTGAGACCCGAAGGCCCTCTG[C/T]CCACTGGCTGCTAGCCTACCTTGCG
Phenotype
MIM: 609106 MIM: 609002
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXO39 PubMed Links

Gene Details

Gene
FBXO39
Gene Name
F-box protein 39
There are no transcripts associated with this gene.

Gene
TEKT1
Gene Name
tektin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053285.1 Intron NP_444515.1
XM_011524027.2 Intron XP_011522329.1

View Full Product Details