Product Details

SNP ID
hCV25929761
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:9587299 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCGTCCCATAAAATACTAACAC[G/T]TATCTCATACTTTCAGCTCGTATGT
Phenotype
MIM: 609009
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
YWHAQ PubMed Links

Gene Details

Gene
YWHAQ
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein theta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006826.3 Intron NP_006817.1

View Full Product Details