Product Details

SNP ID
rs11552473
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:109178269 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTAGATGTCTCCAGTCAGTCGTGT[A/G]GAAATCGATGTGCTTCAGAACAGTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00488 PubMed Links

Gene Details

Gene
LINC00488
Gene Name
long intergenic non-protein coding RNA 488
There are no transcripts associated with this gene.

View Full Product Details