Product Details

SNP ID
rs77827131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49004261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCACAGGAAATCACCTCATGTG[C/T]GCCTCCCACCCACAGGCTTCGCAAG
Phenotype
MIM: 604788
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR6798 PubMed Links

Gene Details

Gene
MIR6798
Gene Name
microRNA 6798
There are no transcripts associated with this gene.

Gene
RUVBL2
Gene Name
RuvB like AAA ATPase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321190.1 642 Silent Mutation TGC,TGT C2C NP_001308119.1
NM_001321191.1 642 Intron NP_001308120.1
NM_006666.2 642 Intron NP_006657.1
XM_011526330.1 642 Intron XP_011524632.1

View Full Product Details