Product Details

SNP ID
rs34708402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:38666308 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCCTAGAGGTCAAAGACAGGCA[A/G]GGGAGAGCCTCGGACGTGGTGCTTG
Phenotype
MIM: 137030
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GALM PubMed Links

Gene Details

Gene
GALM
Gene Name
galactose mutarotase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138801.2 399 Silent Mutation CAA,CAG Q49Q NP_620156.1
XM_011532540.1 399 Silent Mutation CAA,CAG Q49Q XP_011530842.1

View Full Product Details