Product Details

SNP ID
rs35039535
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:63280226 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTGGTAAAAGTCTGTGCTCTCA[C/T]CTGGACCAGCCAAGACCTCATTACT
Phenotype
MIM: 611470
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LGSN PubMed Links

Gene Details

Gene
LGSN
Gene Name
lengsin, lens protein with glutamine synthetase domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143940.1 2476 UTR 3 NP_001137412.1
NM_016571.2 2476 Missense Mutation GAT,GGT D442G NP_057655.2
XM_011535889.2 2476 Missense Mutation GAT,GGT D523G XP_011534191.1
XM_011535892.2 2476 Missense Mutation GAT,GGT D481G XP_011534194.1
XM_017010929.1 2476 Missense Mutation GAT,GGT D523G XP_016866418.1
XM_017010930.1 2476 Missense Mutation GAT,GGT D523G XP_016866419.1
XM_017010931.1 2476 Missense Mutation GAT,GGT D418G XP_016866420.1

View Full Product Details