Product Details
- SNP ID
-
rs61745257
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:58085240 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTCCGGGGCCTTCCCAGGCTGCTC[A/T]TCCTCCTCCTCAGTGGTGCCCGACG
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZSCAN18
PubMed Links
Gene Details
- Gene
- ZSCAN18
- Gene Name
- zinc finger and SCAN domain containing 18
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001145542.1 |
1176 |
Silent Mutation |
GAA,GAT |
E382D |
NP_001139014.1 |
NM_001145543.1 |
1176 |
Silent Mutation |
GAA,GAT |
E326D |
NP_001139015.1 |
NM_001145544.1 |
1176 |
Silent Mutation |
GAA,GAT |
E190D |
NP_001139016.1 |
NM_023926.4 |
1176 |
Silent Mutation |
GAA,GAT |
E326D |
NP_076415.3 |
XM_005259174.4 |
1176 |
Missense Mutation |
GAA,GAT |
E326D |
XP_005259231.1 |
XM_006723335.2 |
1176 |
Missense Mutation |
GAA,GAT |
E326D |
XP_006723398.1 |
XM_011527237.2 |
1176 |
Missense Mutation |
GAA,GAT |
E381D |
XP_011525539.1 |
XM_011527238.1 |
1176 |
Missense Mutation |
GAA,GAT |
E326D |
XP_011525540.1 |
XM_011527239.2 |
1176 |
Missense Mutation |
GAA,GAT |
E326D |
XP_011525541.1 |
XM_017027169.1 |
1176 |
Missense Mutation |
GAA,GAT |
E325D |
XP_016882658.1 |
XM_017027170.1 |
1176 |
Missense Mutation |
GAA,GAT |
E325D |
XP_016882659.1 |
XM_017027171.1 |
1176 |
Silent Mutation |
GAA,GAT |
E325D |
XP_016882660.1 |
View Full Product Details