Product Details

SNP ID
rs6670793
Assay Type
Functionally tested
NCBI dbSNP Submissions
56
Location
Chr.1:27734063 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGTCAGTATTGCAACATAATTGC[A/G]GCATTTATTGGGAATAAATGCCAGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM76A PubMed Links

Gene Details

Gene
FAM76A
Gene Name
family with sequence similarity 76 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143912.1 438 Silent Mutation GCA,GCG A112A NP_001137384.1
NM_001143913.1 438 Silent Mutation GCA,GCG A112A NP_001137385.1
NM_001143914.1 438 Silent Mutation GCA,GCG A78A NP_001137386.1
NM_001143915.1 438 Intron NP_001137387.1
NM_152660.2 438 Silent Mutation GCA,GCG A78A NP_689873.1
XM_017000543.1 438 Silent Mutation GCA,GCG A78A XP_016856032.1

View Full Product Details