Product Details
- SNP ID
-
rs73484047
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:2361301 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTAAAGAACAATTAAGAACTTTAC[A/G]TGGAAAGGGGCCCTTTTCTGCAAAT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ABCA17P
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79821704] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ABCA17P
- Gene Name
- ATP binding cassette subfamily A member 17, pseudogene
There are no transcripts associated with this gene.
View Full Product Details