Product Details

SNP ID
rs72657795
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:37557547 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGAGCAATATACAGGTTGGGGA[C/T]GTGTGGAGCTGAGCTCCTGAACAAT
Phenotype
MIM: 602135 MIM: 609365 MIM: 608241
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNALI1 PubMed Links

Gene Details

Gene
DNALI1
Gene Name
dynein axonemal light intermediate chain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003462.3 Intron NP_003453.2
XM_005271172.2 Intron XP_005271229.1
Gene
GNL2
Gene Name
G protein nucleolar 2
There are no transcripts associated with this gene.

Gene
SNIP1
Gene Name
Smad nuclear interacting protein 1
There are no transcripts associated with this gene.

View Full Product Details