Product Details

SNP ID
hCV25938517
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:71454974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAGGAAGCCTGACTGCATCCCGT[C/T]TGGGAAGGCACAGAGCATTTTTGTT
Phenotype
MIM: 602858 MIM: 608285
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHCR7 PubMed Links

Gene Details

Gene
DHCR7
Gene Name
7-dehydrocholesterol reductase
There are no transcripts associated with this gene.

Gene
NADSYN1
Gene Name
NAD synthetase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018161.4 Intron NP_060631.2

View Full Product Details