Product Details

SNP ID
rs76565267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46019223 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCATGGGAGGGGAGGAATGGGGTG[C/T]GGATCAGCAGGGCCTCAGGGGGAGC
Phenotype
MIM: 604963
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC61 PubMed Links

Gene Details

Gene
CCDC61
Gene Name
coiled-coil domain containing 61
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267723.1 883 Intron NP_001254652.1
XM_005259191.3 883 Intron XP_005259248.1
XM_005259192.4 883 Intron XP_005259249.1
XM_011527257.2 883 Intron XP_011525559.1
Gene
MIR769
Gene Name
microRNA 769
There are no transcripts associated with this gene.

Gene
PGLYRP1
Gene Name
peptidoglycan recognition protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005091.2 883 UTR 3 NP_005082.1

View Full Product Details