Product Details

SNP ID
rs12404886
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:209828198 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGAAGGGGCGTGGCAGGGCTCCCC[A/T]GTCGCCAGAGATGTATCCTCGAGTT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DIEXF PubMed Links

Gene Details

Gene
DIEXF
Gene Name
digestive organ expansion factor homolog (zebrafish)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014388.6 Intron NP_055203.4
XM_006711275.3 Intron XP_006711338.1
XM_011509418.2 Intron XP_011507720.1

View Full Product Details